Learn More
Bcl-2α Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and flow cytometry experiments.
£98.33 - £391.68
Specifications
| Antigen | Bcl-2alpha Ab-1 |
|---|---|
| Clone | 100/D5 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|
12659356
|
Epredia
MS-123-P0 |
100 μL | Concentrated | 200μg/mL | |||||
|
12679356
|
Epredia
MS-123-P1ABX |
100 μL | Concentrated | 1mg/mL | |||||
|
12689356
|
Epredia
MS-123-PABX |
200 μL | Concentrated | 1mg/mL | |||||
|
12669356
|
Epredia
MS-123-P1 |
500 μL | Concentrated | 200μg/mL |
£318.00 £297.48 / 500µL Save £20.52 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
12639356
|
Epredia
MS-123-P |
1 mL | Concentrated | 200μg/mL |
£397.00 £374.32 / 1mL Save £22.68 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
12629366
|
Epredia
MS-123-R7 |
7 mL | Prediluted | Prediluted | |||||
|
12639366
|
Epredia
MS-123-RQ |
12 mL | Ready-to-Use | Prediluted |
£257.00 £240.41 / 12mL Save £16.59 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.Host Species: Mouse
Clone: 100/D5
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse and rat. Others not known.
Epitope: aa 41-54
Immunogen: A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein
Molecular Weight: 25-26kDa
Positive Control: Raji Cells, Tonsil or follicular lymphomas
Cellular Localization: Cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
- Flow Cytometry
Specifications
| Bcl-2alpha Ab-1 | |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| IVD | |
| A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein |
| 100/D5 | |
| Monoclonal | |
| Mouse | |
| Apoptosis | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.