Learn More
Bcl-2α Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and flow cytometry experiments.
Brand: Epredia MS-123-R7
Description
Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.Host Species: Mouse
Clone: 100/D5
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse and rat. Others not known.
Epitope: aa 41-54
Immunogen: A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein
Molecular Weight: 25-26kDa
Positive Control: Raji Cells, Tonsil or follicular lymphomas
Cellular Localization: Cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
- Flow Cytometry
Specifications
| Bcl-2alpha Ab-1 | |
| Monoclonal | |
| Prediluted | |
| Mouse | |
| 7 mL | |
| Apoptosis | |
| Human | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 100/D5 | |
| Unconjugated | |
| A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein | |
| IVD | |
| Primary | |
| Prediluted |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.