missing translation for 'onlineSavingsMsg'
Learn More

Bax Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments.

Brand:  Epredia MS-711-P1

 View more versions of this product

Product Code. 12662097

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Bcl-2, Bax, and Bcl-X are members of a family of proteins that regulate apoptosis. In contrast to Bcl-2α and Bcl-XL, overexpression of bax protein accelerates the programmed cell death (apoptosis).

Host Species: Mouse

Clone: 2D2

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not tested.

Epitope: aa 3-16

Immunogen: A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human Bax protein. This amino acid sequence is not shared by mouse and rat Bax protein.

Molecular Weight: 21kDa

Positive Control: HeLa cells, Hodgkin's lymphoma

Cellular Localization: Cytoplasmic and cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry (Not tested)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bax Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Apoptosis
Human
IgG2a κ
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2D2
Unconjugated
A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein. This amino acid sequence is NOT shared by mouse and rat bax protein.
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.