Learn More
Fisherbrand™ Lab Module for 3 Distributors and 84 Pipettes
Product Code. 17127864 Shop All Fisherbrand Products
Change view
Click to view available options
:
For 3 Distributors and 84 Pipettes
Product Code.
17127864 For 3 Distributors and 84 Pipettes
1 options
Product Code. 17127864
Only null left
Add to basket
Add to basket
Module system creates flexibility in terms of changing to different loads of goods, and helps to avoid heavy lifting, thus ensuring the best possible ergonomic work environment.
  • Glassware module for 84 pipettes and 3 injector units
  • Pipette length of 160 to 630 mm
Specifications
Product Type Lab Module
For Use With (Equipment) 3 Distributors and 84 Pipettes