missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | L3MBTL4 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627295
|
Novus Biologicals
NBP2-39018-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18162199
|
Novus Biologicals
NBP2-39018 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L3MBTL4 Polyclonal specifically detects L3MBTL4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| L3MBTL4 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q8NA19 | |
| 91133 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: IGWCDVTGHPLEVPQRTNDLKILPGQAVCPTPGCRGIGHIRGPRYSGHHSAFGCPYSDMNLKKEATLHDRLREQTQANLESD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ35936, H-l(3)mbt-like protein 4, HsT1031, l(3)mbt-like 4 (Drosophila), L(3)mbt-like protein 4, lethal(3)malignant brain tumor-like protein 4 | |
| L3MBTL4 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title