missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L2HGDH Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £366.00
Specifications
| Antigen | L2HGDH |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18392417
|
Novus Biologicals
NBP3-04541-20UL |
20 μL |
£153.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18301167
|
Novus Biologicals
NBP3-04541-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L2HGDH Polyclonal specifically detects L2HGDH in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| L2HGDH | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 79944 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| 2-hydroxyglutarate dehydrogenase, alpha-ketoglutarate reductase, C14orf160alpha-hydroxyglutarate oxidoreductase, chromosome 14 open reading frame 160, DURANIN, EC 1.1.99.2, FLJ12618, L-2-hydroxyglutarate dehydrogenase, L-2-hydroxyglutarate dehydrogenase, mitochondrial, L-alpha-hydroxyglutarate dehydrogenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human L2HGDH (NP_079160.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title