missing translation for 'onlineSavingsMsg'
Learn More

L2HGDH Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18392417 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392417 20 μL 20µL
18301167 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18392417 Supplier Novus Biologicals Supplier No. NBP30454120UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

L2HGDH Polyclonal specifically detects L2HGDH in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L2HGDH
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 2-hydroxyglutarate dehydrogenase, alpha-ketoglutarate reductase, C14orf160alpha-hydroxyglutarate oxidoreductase, chromosome 14 open reading frame 160, DURANIN, EC 1.1.99.2, FLJ12618, L-2-hydroxyglutarate dehydrogenase, L-2-hydroxyglutarate dehydrogenase, mitochondrial, L-alpha-hydroxyglutarate dehydrogenase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human L2HGDH (NP_079160.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79944
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.