missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-trans-2,4-PDC
GREENER_CHOICE

Product Code. 30001075
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001075

Brand: Tocris Bioscience™ 0298/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Transportable EAAT1-4 inhibitor/non-transportable EAAT5 inhibitor

Potent, competitive, transportable EAAT1-4 inhibitor/non-transportable EAAT5 inhibitor. Also available as part of the Excitatory Amino Acid Transporter Inhibitor Tocriset™ .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64769-66-0
Molecular Formula C6H9NO4

Specifications

Quantity 10 mg
Formula Weight 159.14
Purity >97%
Molecular Weight (g/mol) 159.141
Chemical Name or Material L-trans-2, 4-PDC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.