CAS RN 64769-66-0
CAS RN 64769-66-0
IUPAC Name:
pyrrolidine-2,4-dicarboxylic acid
Synonyms:
pyrrolidine-2,4-dicarboxylic acid
Molecular Weight (g/mol):
159.14
Molecular Formula:
C6H9NO4
InChi Key:
NRSBQSJHFYZIPH-UHFFFAOYNA-N
SMILES:
OC(=O)C1CNC(C1)C(O)=O