Learn More
Masterflex™ L/S™ Standard Pump Head Replacement Rotor Assembly
Product Code. 11815212
Click to view available options
Type:
Replacement Rotor
Unit Size:
Each
Product Code. 11815212
Brand: Masterflex™ 0701395
Only null left
Add to basket
Add to basket
Masterflex™ L/S™ Standard Pump Head Replacement Rotor Assembly for use with I/P™ Standard Pump Heads
  • Choose replacement parts suitable for your I/P™ Standard pump head size
  • Replacement parts to keep pump heads operating for an extended period of time
  • Two end bell assemblies are required for a complete head assembly
Specifications
For Use With L/S™ Standard Pump Heads 07013-21, 07013-52, 07014-21, 07014-52, 07016-21, 07016-52, 07017-21, 07017-52, 07018-21, and 07018-52
Type Replacement Rotor