Learn More
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assembly
Click to view available options
For Use With:
CRS Rotor, L/S 14 Tubing
CRS Rotor, L/S 15 Tubing
CRS Rotor, L/S 16 Tubing
CRS Rotor, L/S 18 Tubing
CRS Rotor, L/S 24 Tubing
SS Rotor, L/S 14 Tubing
Unit Size:
Each
Description
- Replacement parts to keep pump head operating for an extended period of time
- Two end bell assemblies are required for a complete head assembly
- Choose replacement parts suitable for your I/P™ Standard pump head size
- Suitable for use with L/S™ Standard Pump Heads
Specifications
Specifications
| For Use With | CRS Rotor, L/S 15 Tubing |
| Material | Polycarbonate |
| Size | L/S™ Standard Pump Heads 07015-00, 07015-20, 07015-70, and 07015-72 |
| Tubing Size | L/S 15 |
| Type | Replacement End Bell Assembly |