missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ and I/P™ Digital Modular Drives Extension Cable

Product Code. 15498140
Click to view available options
Overall Length (English):
9 ft.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15498140

Brand: Masterflex™ 7730182

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Masterflex™ L/S™ and I/P™ Digital Modular Drives Extension Cable increases modular drive flexibility by 9 ft.

  • Easily connect to Masterflex™ pumps with connectors on both ends
  • Masterflex extension cables can be used with L/S and I/P modular drives to extend the distance between drive motor and controller
  • Cable is 9 ft (2.7 m) long to increase installation flexibility
  • Extension cable is compatible with L/S™ pump models 77301-40, -50, -60, and -70; and with I/P™ pump models 07594-00 and -10

Specifications

For Use With (Application) Distance Extension
For Use With L/S™ Pump Models: 77301-40, -50, -50, and -70. I/P™ Pump Models: 07594-00, and -10.
Overall Length (English) 9 ft.
Overall Length (Metric) 2.7 m
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.