Learn More
Masterflex™ L/S™ High-Performance Pump Head Tubing Retainer Kit
Click to view available options
Type:
Service Kit
Unit Size:
Each
Description
- Tubing retainer prevents tubing from moving within the pump head
- This service kit extends the life of Masterflex pump heads
- Compatible with the Masterflex™ L/S™ High-Performance Pump Heads 77250-62
Specifications
Specifications
| For Use With (Application) | Tubing retainer prevents tubing from moving within the pump head |
| For Use With | L/S™ High-Performance Pump Heads (77250-62) |
| Type | Service Kit |