Learn More
Masterflex™ L/S™ Easy-Load™ Replacement Rotor and Bearing Support Assembly
Product Code. 15550924
Click to view available options
For Use With:
L/S Easy-Load Pump Head 07018-60
Unit Size:
Each
Product Code. 15550924
Brand: Masterflex™ 0751868
Only null left
Add to basket
Add to basket
Masterflex™ L/S™ Easy-Load™ Replacement Rotor and Bearing Support Assembly for I/P™ Standard Pump Heads
  • Extend the life of Masterflex™ I/P™ Standard Pump Heads with these replacement parts
  • Choose replacement parts suitable for your I/P™ standard pump head size
  • Suitable for Easy-Load™ Pump Head 07518-60
  • Two end bell assemblies are required for a complete head assembly
Specifications
For Use With L/S Easy-Load Pump Head 07018-60
Material Polyphenylene Sulfide
Tubing Type L/S™
Type Replacement Rotor and Bearing Support Assembly
Rotor Material Stainless Steel