missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine ethyl ester dihydrochloride, 99%

Product Code. 11402858
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11402858

Brand: Thermo Scientific Alfa Aesar A16824.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3844-53-9 | C8H20Cl2N2O2 | 247.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3844-53-9
Molecular Formula C8H20Cl2N2O2
Molecular Weight (g/mol) 247.16
MDL Number MFCD00039068
InChI Key DZIYAIZKJOHVQC-KLXURFKVSA-N
Synonym l-lysine ethyl ester dihydrochloride, h-lys-oet.2hcl, ethyl l-lysinate dihydrochloride, ethyl 2s-2,6-diaminohexanoate dihydrochloride, ethyl l-lysinate hcl, s-ethyl 2,6-diaminohexanoate dihydrochloride, l-lysine, ethyl ester, dihydrochloride, h-lys-oet·2hcl, h-lys-oet inverted exclamation mark currency2hcl, lys-oet2hcl
PubChem CID 107468
IUPAC Name ethyl (2S)-2,6-diaminohexanoate;dihydrochloride
SMILES Cl.Cl.CCOC(=O)[C@@H](N)CCCCN

Specifications

Melting Point ∼145°C (decomposition)
Beilstein 3697099
Quantity 100 g
Formula Weight 247.17
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Lysine ethyl ester dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.