accessibility menu, dialog, popup

UserName

OONH2(S)H2NHClHClCH3

CAS RN 3844-53-9

IUPAC Name: hydrogen ethyl 2,6-diaminohexanoate chloride
Synonyms: hydrogen ethyl 2,6-diaminohexanoate chloride
Molecular Weight (g/mol): 210.7
Molecular Formula: C8H19ClN2O2
InChi Key: SGEJVUZMEHMANQ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCOC(=O)C(N)CCCCN
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)