missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine tert-butyl ester hydrochloride, 98%

Product Code. 15486438
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15486438 1 g 1g
15496438 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15486438

Brand: Thermo Scientific Alfa Aesar H59980.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2748-02-9 | C3H10NO | 76.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2748-02-9
MDL Number MFCD00038906

Specifications

Molecular Formula C10H22ClNO2
Beilstein 3697141
Synonym h-leu-otbu.hcl, l-leucine tert-butyl ester hydrochloride, h-leu-otbu hcl, l-leucine t-butyl ether hcl salt, l-leucine t-butyl ester hydrochloride, l-leucine tert-butyl ester hcl, tert-butyl l-leucinate hydrochloride, s-tert-butyl 2-amino-4-methylpentanoate hydrochloride, tert-butyl 2s-2-amino-4-methylpentanoate hydrochloride, h-leu-obut hcl
InChI Key RFUWRXIYTQGFGA-QRPNPIFTSA-N
SMILES Cl.CC(C)C[C@H](N)C(=O)OC(C)(C)C
IUPAC Name tert-butyl (2S)-2-amino-4-methylpentanoate;hydrochloride
Molecular Weight (g/mol) 223.74
Quantity 1 g
PubChem CID 12451443
Formula Weight 223.74
Optical Rotation +12° (c=2 in DMF)
Percent Purity 98%
Chemical Name or Material L-Leucine tert-butyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.