CAS RN 2748-02-9
CAS RN 2748-02-9
IUPAC Name:
hydrogen tert-butyl (2S)-2-amino-4-methylpentanoate chloride
Synonyms:
hydrogen tert-butyl (2S)-2-amino-4-methylpentanoate chloride
Molecular Weight (g/mol):
223.74
Molecular Formula:
C10H22ClNO2
InChi Key:
RFUWRXIYTQGFGA-QRPNPIFTSA-N
SMILES:
[H+].[Cl-].CC(C)C[C@H](N)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
L-Leucine tert-butyl ester hydrochloride, 98%
CAS: 2748-02-9 MDL Number: MFCD00038906 ChEBI: CHEBI:78502