missing translation for 'onlineSavingsMsg'
Learn More

L-gamma-Glutamyl-L-valine, TRC

Product Code. 30401678
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401678 50 mg -
30407966 100 mg -
30392444 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401678 Supplier Toronto Research Chemicals Supplier No. TRCH25381050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2746-34-1
Molecular Formula C10 H18 N2 O5
Molecular Weight (g/mol) 246.26
Synonym N-L-γ-Glutamyl-L-valine, N-L-γ-Glutamyl-(6CI,7CI) Valine, N-L-γ-Glutamyl-L-(8CI) Valine
IUPAC Name (2S)-2-amino-5-[[(1S)-1-carboxy-2-methylpropyl]amino]-5-oxopentanoic acid
SMILES CC(C)[C@H](NC(=O)CC[C@H](N)C(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C10H18N2O5/c1-5(2)8(10(16)17)12-7(13)4-3-6(11)9(14)15/h5-6,8H,3-4,11H2,1-2H3,(H,12,13)(H,14,15)(H,16,17)/t6-,8-/m0/s1
Quantity 50 mg
Formula Weight 246.122
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-gamma-Glutamyl-L-valine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.