CAS RN 2746-34-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-gamma-Glutamyl-L-valine, TRC
CAS: 2746-34-1 Molecular Formula: C10 H18 N2 O5 Molecular Weight (g/mol): 246.26 Synonym: N-L-γ-Glutamyl-L-valine,N-L-γ-Glutamyl-(6CI,7CI) Valine,N-L-γ-Glutamyl-L-(8CI) Valine IUPAC Name: (2S)-2-amino-5-[[(1S)-1-carboxy-2-methylpropyl]amino]-5-oxopentanoic acid SMILES: CC(C)[C@H](NC(=O)CC[C@H](N)C(=O)O)C(=O)O