missing translation for 'onlineSavingsMsg'
Learn More

L-Fucitol, TRC

Product Code. 30407863
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407863 10 mg
30389101 5 mg
30389827 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407863 Supplier Toronto Research Chemicals Supplier No. TRCF83614310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13074-06-1
Molecular Formula C6H14O5
Molecular Weight (g/mol) 166.17
SMILES C[C@H](O)[C@@H](O)[C@@H](O)[C@H](O)CO

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C6H14O5/c1-3(8)5(10)6(11)4(9)2-7/h3-11H,2H2,1H3/t3-,4+,5+,6-/m0/s1
Formula Weight 166.0841
Chemical Name or Material L-Fucitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.