CAS RN 13074-06-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-Fucitol, TRC
CAS: 13074-06-1 Molecular Formula: C6H14O5 Molecular Weight (g/mol): 166.17 SMILES: C[C@H](O)[C@@H](O)[C@@H](O)[C@H](O)CO