missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid 6-Stearate, TRC

Product Code. 30008267
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008267 50 mg 50mg
30008181 250 mg 250mg
30006733 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008267 Supplier Toronto Research Chemicals Supplier No. TRCA78698550MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10605-09-1
Molecular Formula C24H42O7
Synonym Ascorbyl 6-stearate∣6-O-Stearoyl-L-ascorbic Acid∣Vitamin C monostearate∣L-Ascorbic acid monostearate;

Specifications

Quantity 50 mg
Shipping Condition Room Temperature
Formula Weight 442.293
Molecular Weight (g/mol) 442.59
InChI Key NRHBEGWPKVFGRH-UHFFFAOYNA-N
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
IUPAC Name 2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl octadecanoate
Chemical Name or Material Ascorbyl Stearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.