CAS RN 10605-09-1
IUPAC Name:
2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl octadecanoate
Synonyms:
ascorbyl stearate
Molecular Weight (g/mol):
442.59
Molecular Formula:
C24H42O7
InChi Key:
NRHBEGWPKVFGRH-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
1
–
1
of
1
results
1
L-Ascorbic Acid 6-Stearate, TRC
CAS: 10605-09-1 Molecular Formula: C24H42O7 Synonym: Ascorbyl 6-stearate∣6-O-Stearoyl-L-ascorbic Acid∣Vitamin C monostearate∣L-Ascorbic acid monostearate;