missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-AP5, Tocris Bioscience™
Potent and selective NMDA antagonist; less active form of DL-AP5
£168.21 - £558.87
Chemical Identifiers
CAS | 79055-67-7 |
---|---|
Molecular Formula | C5H12NO5P |
Molecular Weight (g/mol) | 197.127 |
InChI Key | VOROEQBFPPIACJ-BYPYZUCNSA-N |
Synonym | l-ap5, s-2-amino-5-phosphonopentanoic acid, 2s-2-amino-5-phosphonopentanoic acid, chembl71898, 5-phosphono-l-norvaline, tocris-0107, s-2-amino-5-phosphonovaleric acid, s-2-amino-5-phosphonopentanoicacid, s-2-amino-5-phosphono-pentanoic acid, l-+-2-amino-5-phosphonopentanoic acid |
PubChem CID | 1617110 |
IUPAC Name | (2S)-2-amino-5-phosphonopentanoic acid |
SMILES | C(CC(C(=O)O)N)CP(=O)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18094242
|
Tocris Bioscience
0107/10 |
10 mg |
£168.21
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18035542
|
Tocris Bioscience
0107/50 |
50 mg |
£558.87
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NMDA antagonist. Also agonist at quisqualate-sensitized AP6 site, where it is more potent than the isomer D-AP5 .DL Mixture and D-isomer also available.
Chemical Identifiers
79055-67-7 | |
197.127 | |
l-ap5, s-2-amino-5-phosphonopentanoic acid, 2s-2-amino-5-phosphonopentanoic acid, chembl71898, 5-phosphono-l-norvaline, tocris-0107, s-2-amino-5-phosphonovaleric acid, s-2-amino-5-phosphonopentanoicacid, s-2-amino-5-phosphono-pentanoic acid, l-+-2-amino-5-phosphonopentanoic acid | |
(2S)-2-amino-5-phosphonopentanoic acid |
C5H12NO5P | |
VOROEQBFPPIACJ-BYPYZUCNSA-N | |
1617110 | |
C(CC(C(=O)O)N)CP(=O)(O)O |
Specifications
79055-67-7 | |
l-ap5, s-2-amino-5-phosphonopentanoic acid, 2s-2-amino-5-phosphonopentanoic acid, chembl71898, 5-phosphono-l-norvaline, tocris-0107, s-2-amino-5-phosphonovaleric acid, s-2-amino-5-phosphonopentanoicacid, s-2-amino-5-phosphono-pentanoic acid, l-+-2-amino-5-phosphonopentanoic acid | |
C(CC(C(=O)O)N)CP(=O)(O)O | |
197.127 | |
197.13 | |
L-AP5 |
C5H12NO5P | |
VOROEQBFPPIACJ-BYPYZUCNSA-N | |
(2S)-2-amino-5-phosphonopentanoic acid | |
1617110 | |
>99% |
Safety and Handling
Recommended Storage : Store at Room Temperature