missing translation for 'onlineSavingsMsg'
Learn More

L-AP5, Tocris Bioscience™

Catalog Number 18094242
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
79055-67-7
C5H12NO5P
197.127
VOROEQBFPPIACJ-BYPYZUCNSA-N
l-ap5, s-2-amino-5-phosphonopentanoic acid, 2s-2-amino-5-phosphonopentanoic acid, chembl71898, 5-phosphono-l-norvaline, tocris-0107, s-2-amino-5-phosphonovaleric acid, s-2-amino-5-phosphonopentanoicacid, s-2-amino-5-phosphono-pentanoic acid, l-+-2-amino-5-phosphonopentanoic acid
1617110
(2S)-2-amino-5-phosphonopentanoic acid
C(CC(C(=O)O)N)CP(=O)(O)O
This item is not returnable. View return policy
79055-67-7
C5H12NO5P
197.127
VOROEQBFPPIACJ-BYPYZUCNSA-N
l-ap5, s-2-amino-5-phosphonopentanoic acid, 2s-2-amino-5-phosphonopentanoic acid, chembl71898, 5-phosphono-l-norvaline, tocris-0107, s-2-amino-5-phosphonovaleric acid, s-2-amino-5-phosphonopentanoicacid, s-2-amino-5-phosphono-pentanoic acid, l-+-2-amino-5-phosphonopentanoic acid
1617110
(2S)-2-amino-5-phosphonopentanoic acid
C(CC(C(=O)O)N)CP(=O)(O)O

Potent and selective NMDA antagonist; less active form of DL-AP5

NMDA antagonist. Also agonist at quisqualate-sensitized AP6 site, where it is more potent than the isomer D-AP5 .

DL Mixture and D-isomer also available.

Quantity 10 mg
Formula Weight 197.13
Percent Purity >99%
Chemical Name or Material L-AP5
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'