missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-655,708

Product Code. 18015292
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18015292

Brand: Tocris Bioscience™ 1327/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Benzodiazepine inverse agonist; selective for α5-containing GABAA receptors

Potent, selective inverse agonist for the benzodiazepine site of GABAA receptors containing the α5 subunit (Ki = 0.45 nM). Displays 50-100-fold selectivity over GABAA receptors containing α1, α2, α3 or α6 subunits in combination with β3 and γ2. Enhances LTP in a mouse hippocampal slice model and increases spatial learning, without displaying proconvulsant activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130477-52-0
Molecular Formula C18H19N3O4

Specifications

Quantity 10 mg
Formula Weight 341.37
Purity >99%
Molecular Weight (g/mol) 341.367
Chemical Name or Material L-655, 708
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.