Learn More
L(-)-2-Octanol, 99+%, Thermo Scientific Chemicals
£23.60 - £126.00
Chemical Identifiers
CAS | 5978-70-1 |
---|---|
Molecular Formula | C8H18O |
Molecular Weight (g/mol) | 130.23 |
MDL Number | MFCD00064284 |
InChI Key | SJWFXCIHNDVPSH-MRVPVSSYSA-N |
Synonym | r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol |
PubChem CID | 80080 |
ChEBI | CHEBI:37871 |
IUPAC Name | (2R)-octan-2-ol |
SMILES | CCCCCCC(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10311084
|
Thermo Scientific Acros
164590010 |
1 g | Glass bottle |
£23.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10571991
|
Thermo Scientific Acros
164590050 |
5 g | Glass bottle |
£75.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10720671
|
Thermo Scientific Acros
164590100 |
10 g | Glass bottle |
£126.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5978-70-1 | |
130.23 | |
SJWFXCIHNDVPSH-MRVPVSSYSA-N | |
80080 | |
(2R)-octan-2-ol |
C8H18O | |
MFCD00064284 | |
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol | |
CHEBI:37871 | |
CCCCCCC(C)O |
Specifications
5978-70-1 | |
0.8300g/mL | |
60°C | |
99% min. (GC) | |
CH3(CH2)5CH(OH)CH3 | |
MFCD00064284 | |
15, 6841 | |
−9.50° (20°C neat) | |
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol | |
SJWFXCIHNDVPSH-MRVPVSSYSA-N | |
(2R)-octan-2-ol | |
80080 | |
130.23 | |
Liquid |
Colorless to Yellow | |
175.0°C | |
Authentic | |
C8H18O | |
1.4250 to 1.4270 | |
01, IV, 1770 | |
0.83 | |
− 9.50 | |
Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol and ether | |
CCCCCCC(C)O | |
130.23 | |
CHEBI:37871 | |
99+% | |
L(-)-2-Octanol, 99+% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 227-777-0
missing translation for 'tsca' : TSCA
- L(-)-2-Octanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only