missing translation for 'onlineSavingsMsg'
Learn More

L(-)-2-Octanol, 99+%

Product Code. 10311084 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10311084 1 g Glass bottle 1g
10571991 5 g Glass bottle 5g
10720671 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10311084 Supplier Thermo Scientific Acros Supplier No. 164590010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5978-70-1 | C8H18O | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5978-70-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00064284
InChI Key SJWFXCIHNDVPSH-MRVPVSSYSA-N
Synonym r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol
PubChem CID 80080
ChEBI CHEBI:37871
IUPAC Name (2R)-octan-2-ol
SMILES CCCCCCC(C)O

Specifications

Color Colorless to Yellow
Density 0.8300g/mL
Boiling Point 175.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5CH(OH)CH3
Refractive Index 1.4250 to 1.4270
Quantity 1 g
Beilstein 01, IV, 1770
Merck Index 15, 6841
Specific Gravity 0.83
Specific Rotation Condition −9.50° (20°C neat)
Specific Rotation − 9.50
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol and ether
Formula Weight 130.23
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material L(-)-2-Octanol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • L(-)-2-Octanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.