missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv8.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£433.00
Specifications
| Antigen | Kv8.1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Kv8.1 Polyclonal specifically detects Kv8.1 in Human samples. It is validated for Western Blot.Specifications
| Kv8.1 | |
| Polyclonal | |
| Rabbit | |
| Q6PIU1 | |
| 27012 | |
| IgG | |
| 55 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| HNKA, KCNB3, KV2.3, KV8.1, Neuronal potassium channel alpha subunit HNKA, potassium channel Kv8.1, potassium channel, subfamily V, member 1, potassium voltage-gated channel subfamily V member 1, Voltage-gated potassium channel subunit Kv8.1 | |
| KCNV1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title