missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv8.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37847
This item is not returnable.
View return policy
Description
Kv8.1 Polyclonal specifically detects Kv8.1 in Human samples. It is validated for Western Blot.
Specifications
| Kv8.1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| HNKA, KCNB3, KV2.3, KV8.1, Neuronal potassium channel alpha subunit HNKA, potassium channel Kv8.1, potassium channel, subfamily V, member 1, potassium voltage-gated channel subfamily V member 1, Voltage-gated potassium channel subunit Kv8.1 | |
| Rabbit | |
| 55 kDa | |
| 100 μL | |
| Signal Transduction | |
| 27012 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q6PIU1 | |
| KCNV1 | |
| The immunogen for Anti-KCNV1 antibody is: synthetic peptide directed towards the N-terminal region of Human KCNV1.. Peptide sequence: LLDSPLDSGSLTSLDSSVFCSEGEGEPLALGDCFTVNVGGSRFVLSQQAL | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Canine, Equine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction