missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KOR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £310.00
Specifications
| Antigen | KOR |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18270775
|
Novus Biologicals
NB110-79876SS |
0.025 mg |
£154.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18286750
|
Novus Biologicals
NB110-79876 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KOR Polyclonal specifically detects KOR in Human, Rat samples. It is validated for Western Blot.Specifications
| KOR | |
| Western Blot 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| GPCR, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| 4986 | |
| Synthetic peptide comprising residues 366-380 of the human KOR-1 protein. Sequence identity is 66% in mouse and rat. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 50 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK | |
| OPRK1 | |
| IgG | |
| Affinity Purified | |
| Kappa Opioid Receptor |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title