missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KOR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-79876SS
This item is not returnable.
View return policy
Description
KOR Polyclonal specifically detects KOR in Human, Rat samples. It is validated for Western Blot.
Specifications
| KOR | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| OPRK1 | |
| Synthetic peptide comprising residues 366-380 of the human KOR-1 protein. Sequence identity is 66% in mouse and rat. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Kappa Opioid Receptor | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000 | |
| kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK | |
| Rabbit | |
| 50 kDa | |
| 0.025 mg | |
| GPCR, Signal Transduction | |
| 4986 | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction