missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Reaction-Jacketed Beakers

Product Code. p-3206692
Click to view available options
Capacity (Metric):
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 16600603

Brand: DWK Life Sciences 3170000100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The open top provides easy access to concentrated residual samples.

  • A conical lower interior surface eliminates spin-out when used with an egg-shaped PTFE magnetic stir bar
  • Jacket has serrated 3/8 in. hose connections
  • Design meets ASTM Specification E960, Type I requirements
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 3.38 oz.
Description Connection: 0.37 in. Serrated Hose
Diameter (English) Inner 1.85 in.
Diameter (English) Outer 3.14 in.
Diameter (Metric) Inner 4.7 cm
Diameter (Metric) Outer 8 cm
Capacity (Metric) 100 mL
Material Borosilicate Glass
For Use With (Application) Laboratory Applications Requiring a Variety of Temperature-Controlled Reactions and Useful in Biological and Chemical Reactions Including Titrations, Enzyme Digestions and Synthetic Organic Reactions
Height (English) Inner 2.87 in.
Height (English) Outer 3.34 in.
Height (Metric) Inner 7.3 cm
Height (Metric) Outer 8.5 cm
Includes Conical Lower Interior Surface
Type Beaker
Certifications/Compliance ASTM E960, ASTM E438, USP Type I
Style Open Top
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.