missing translation for 'onlineSavingsMsg'
Learn More

KMT1A/SUV39H1 Antibody (44.1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

£184.00

Specifications

Antigen KMT1A/SUV39H1
Clone 44.1
Concentration 2 mg/mL
Dilution Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Applications Western Blot
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18591241
View Documents
Novus Biologicals
NB120-12405-0.025ml
0.025 mL
£184.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KMT1A/SUV39H1 Monoclonal specifically detects KMT1A/SUV39H1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

KMT1A/SUV39H1
2 mg/mL
Western Blot
Unconjugated
Mouse
Chromatin Research
H3-K9-HMTase 1, Histone H3-K9 methyltransferase 1, histone-lysine N-methyltransferase SUV39H1, histone-lysine N-methyltransferase, H3 lysine-9 specific 1, KMT1AEC 2.1.1.43, Lysine N-methyltransferase 1A, MG44, Position-effect variegation 3-9 homolog, Su(var)3-9 homolog 1, suppressor of variegation 3-9 (Drosophila) homolog 1, Suppressor of variegation 3-9 homolog 1, suppressor of variegation 3-9 homolog 1 (Drosophila), Suppressor of variegation 3-9 homolog 1, Suvar) 3-9 homolog, SUV39H
SUV39H1
IgG1
Protein A or G purified
Monoclonal Anti-SUV39H1 Histone Methyltransferase recognizes an epitope in the N-terminal (195 amino acids) of human and mouse SUV39H1 Histone Methyltransferase.
44.1
Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Monoclonal
Purified
RUO
Human
6839
Recombinant fusion protein, MBP-SUV39H1.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.