missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT1A/SUV39H1 Antibody (44.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB120-12405-0.025ml
This item is not returnable.
View return policy
Description
KMT1A/SUV39H1 Monoclonal specifically detects KMT1A/SUV39H1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| KMT1A/SUV39H1 | |
| Monoclonal | |
| 2 mg/mL | |
| Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
| SUV39H1 | |
| Recombinant fusion protein, MBP-SUV39H1. | |
| 0.025 mL | |
| Chromatin Research | |
| 6839 | |
| Human | |
| Purified |
| Western Blot | |
| 44.1 | |
| Unconjugated | |
| H3-K9-HMTase 1, Histone H3-K9 methyltransferase 1, histone-lysine N-methyltransferase SUV39H1, histone-lysine N-methyltransferase, H3 lysine-9 specific 1, KMT1AEC 2.1.1.43, Lysine N-methyltransferase 1A, MG44, Position-effect variegation 3-9 homolog, Su(var)3-9 homolog 1, suppressor of variegation 3-9 (Drosophila) homolog 1, Suppressor of variegation 3-9 homolog 1, suppressor of variegation 3-9 homolog 1 (Drosophila), Suppressor of variegation 3-9 homolog 1, Suvar) 3-9 homolog, SUV39H | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Monoclonal Anti-SUV39H1 Histone Methyltransferase recognizes an epitope in the N-terminal (195 amino acids) of human and mouse SUV39H1 Histone Methyltransferase. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction