missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMO Mouse, anti-Human, CoraLite 594, Clone: 2D2F2, Proteintech
Mouse Monoclonal Antibody
£159.65 - £416.33
Specifications
| Antigen | KMO |
|---|---|
| Clone | 2D2F2 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite 594 |
Description
The KMO antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KMO. This antibody recognizes human, mouse, rat antigen. The KMO antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| KMO | |
| 1000 μg/mL | |
| CoraLite 594 | |
| Mouse | |
| 8564 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2D2F2 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| dJ317G22.1, KMO, Kynurenine 3 hydroxylase, Kynurenine 3 monooxygenase | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title