Learn More
KMO Mouse, anti-Human, CoraLite 594, Clone: 2D2F2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60029-20UL
Description
The KMO antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KMO. This antibody recognizes human, mouse, rat antigen. The KMO antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| KMO | |
| 2D2F2 | |
| CoraLite 594 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| dJ317G22.1, KMO, Kynurenine 3 hydroxylase, Kynurenine 3 monooxygenase | |
| 20 μL | |
| 8564 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |