missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRA1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | KLRA1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 4-20 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
KLRA1 Polyclonal specifically detects KLRA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| KLRA1 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 4-20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| EAW96194 | |
| 10748 | |
| Antibody was raised against a 13 amino acid synthetic peptide near the center of human KLRA1. The immunogen is located within amino acids 140 - 190 of KLRA1. Amino Acid Squence: CYYFTMQKKDWKG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| killer cell lectin-like receptor subfamily A pseudogene 1, KLRA#, KLRA1, Ly49, Ly-49L, LY49L, MGC126520, MGC126522 | |
| KLRA1P | |
| IgG | |
| Affinity Purified | |
| At least three isoforms of KLRA1 are known to exist; this antibody recognizes only the long isoform. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title