missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRA1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77343-0.025mg
This item is not returnable.
View return policy
Description
KLRA1 Polyclonal specifically detects KLRA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| KLRA1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| killer cell lectin-like receptor subfamily A pseudogene 1, KLRA#, KLRA1, Ly49, Ly-49L, LY49L, MGC126520, MGC126522 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10748 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 4-20 μg/mL | |
| EAW96194 | |
| KLRA1P | |
| Antibody was raised against a 13 amino acid synthetic peptide near the center of human KLRA1. The immunogen is located within amino acids 140 - 190 of KLRA1. Amino Acid Squence: CYYFTMQKKDWKG | |
| 0.025 mg | |
| Primary | |
| At least three isoforms of KLRA1 are known to exist; this antibody recognizes only the long isoform. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction