missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Klotho beta Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
£187.00 - £504.00
Specifications
| Antigen | Klotho beta |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18748298
![]() |
R&D Systems
AF2619-SP |
25 μg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18048491
|
R&D Systems
AF2619 |
100 μg |
£504.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Klotho beta Polyclonal specifically detects Klotho beta in Mouse samples. It is validated for Western Blot.Specifications
| Klotho beta | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
| KLB | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_112457 | |
| 152831 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Klotho beta (R&D Systems, Catalog # 2619-KB) Phe53-Leu995 Accession # NP_112457 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Klotho beta in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant mouse Klotho is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title