missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Klotho beta Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF2619
This item is not returnable.
View return policy
Description
Klotho beta Polyclonal specifically detects Klotho beta in Mouse samples. It is validated for Western Blot.
Specifications
| Klotho beta | |
| Polyclonal | |
| Unconjugated | |
| NP_112457 | |
| KLB | |
| Mouse myeloma cell line NS0-derived recombinant mouse Klotho beta (R&D Systems, Catalog # 2619-KB) Phe53-Leu995 Accession # NP_112457 | |
| 100 μg | |
| Primary | |
| Detects mouse Klotho beta in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant mouse Klotho is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 152831 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction