missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Kligler Iron Slant Agar With Urea
Description
Kligler Iron Slant Agar With Urea is used to differentiate enteric bacteria based on their ability to ferment glucose and lactose, produce hydrogen sulfide (H2S), and hydrolyze urea, aiding in their identification.
Specifications
Specifications
| Color | Red Orange, Translucent |
| Content And Storage | 2°C to 25°C |
| Format | Poured Tube, Slant Agar |
| Manufacturing Origin | Germany |
| Product Type | Agar |
| Fill Weight | 8.5 to 9.5 g |
| Packaging Type | 50 Tubes/Box |
| Quantity | 50/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?