missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £274.00
Specifications
| Antigen | KLF14 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18339942
|
Bio-Techne
NBP3-15576-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18316934
|
Novus Biologicals
NBP3-15576-100UL |
100 μg |
£274.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLF14 Polyclonal antibody specifically detects KLF14 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| KLF14 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 136259 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| basic transcription element-binding protein 5, Kruppel-like factor 14 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human KLF14 (NP_619638.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title