missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-15576-100UL
This item is not returnable.
View return policy
Description
KLF14 Polyclonal antibody specifically detects KLF14 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| KLF14 | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 136259 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| basic transcription element-binding protein 5, Kruppel-like factor 14 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human KLF14 (NP_619638.2). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction