missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ A50 Breathable Splash and Particle Protection Hooded Coveralls, White

Product Code. 11720684
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
Pack of 25
This item is not returnable. View return policy

Product Code. 11720684

Brand: KleenGuard™ 96820

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Short-life, breathable hooded coveralls with storm flap for respirator. KleenGuard A50 Breathable Splash and Particle Protection Hooded Coveralls are specially designed for comfort, safety and strength.

  • White, limited life hooded coverall
  • Supplied in packs of 25
  • Provides PPE Category III protection
  • Storm flap and hood designed for easier taping to a respirator
  • Provides anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
  • Durable SMS fabric with additional treatment for increased protection
  • Available in sizes Small through 3X-Large

Specifications

Product Type A50 Breathable Particle and Splash Protection Coveralls
Size Medium
Gender Unisex
Material Anti-static SMS
Color White
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type 2-way Zipper
Seam Type Stitched
Sterility Non-sterile
Protection Class A50 PPE Category III
Resistance Perforation, Flame, Abrasion, Tears, Particle, Liquid
Length (English) 23.4 in.
Length (Metric) 60 cm
Weight (English) 12.001 lb.
Weight (Metric) 5.455 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.