Learn More
Invitrogen™ KIT [T670I] Recombinant Human Protein
KIT T670I is a Gleevec resistant form of KIT.
Brand: Invitrogen™ PR7423A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
KIT is a proto-oncogene encoding a transmembrane tyrosine kinase receptor for stem cell factor required for normal hematopoiesis, melangenesis and gametogenesis.
Specifications
NP_000213, T670I | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
KIT T670I, cKit, PBT | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
53.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
PDGFR Kinase Family | |
Recombinant | |
Baculovirus |