missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIT [T670I] Recombinant Human Protein
KIT T670I is a Gleevec resistant form of KIT.
£2555.00 - £12060.00
Specifications
Accession Number | NP_000213, T670I |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 53.3kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468583
|
Invitrogen™
PR7423A |
100μg |
£2555.00
100µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10221073
|
Invitrogen™
PV4187 |
1mg |
£12060.00
1mg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
KIT is a proto-oncogene encoding a transmembrane tyrosine kinase receptor for stem cell factor required for normal hematopoiesis, melangenesis and gametogenesis.
Specifications
NP_000213, T670I | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
PDGFR Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
53.3kDa | |
RUO - research use only | |
KIT T670I, cKit, PBT | |
Human | |
His Tag | |
Frozen |