missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KIT, His Tag Recombinant Protein
KIT Recombinant Human Protein
£438.00 - £6900.00
Specifications
Accession Number | NP_000213 |
---|---|
Gene Alias | KIT, cKit, PBT |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11579286
|
Invitrogen™
P3081 |
10μg |
£438.00
10µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10083172
|
Invitrogen™
PV3589 |
1mg |
£6900.00
1mg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15458403
|
Invitrogen™
PR4460D |
100 μg |
£1602.00
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
KIT is a proto-oncogene encoding a transmembrane tyrosine kinase receptor for stem cell factor required for normal hematopoiesis, melangenesis and gametogenesis.Specifications
NP_000213 | |
Human | |
Baculovirus |
KIT, cKit, PBT | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.