missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KIT, His Tag Recombinant Protein
KIT Recombinant Human Protein
Brand: Invitrogen™ PR4460D
Code : NEW
Additional Details : Weight : 0.01000kg
Description
KIT is required for normal hematopoiesis, melangenesis and gametogenesis.
Specifications
NP_000213 | |
53.3 kDa | |
KIT, cKit, PBT | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Recombinant, Cytoplasmic | |
Oncology, Signal Transduction |
Kinase Assay | |
100 μg | |
PDGFR Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 100mM NaCl, 0.1mM EDTA, 0.05% NP-40, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.